Learn More
Bio Rad Laboratories HYDROXYAPATITE CHT CERAMIC
Supplier:  Bio Rad Laboratories 1584000
Encompass
Ceramic Hydroxyapatite; Bio-Rad; CHT; Spherical; Macroporous form; Used reproducibly for several hundred cycles at high flow rates and in large columns; Type I; 40um particle size; 10g
Catalog No. 50-901-3165