Learn More
Bio Rad Laboratories HYDROXYAPATITE CHT CERAMIC
Catalog No. 509010238
Encompass
Hydroxyapatite, Ceramic; Bio-Rad; CHT; Type I; 80um particle size; 100g
Catalog No. 50-901-0238 Supplier Bio Rad Laboratories Supplier No. 1570080
Hydroxyapatite, Ceramic; Bio-Rad; CHT; Type I; 80um particle size; 100g