Learn More
Bio Rad Laboratories HYDROXYAPATITE CHT CERAMIC
Supplier:  Bio Rad Laboratories 1570040
Encompass
Hydroxyapatite; Bio-Rad; CHT ceramic; Type I; High protein binding capacity (> 25mg of lysozyme/g); Higher capacity for acidic proteins; 40um particle size; 100g
Catalog No. 50-901-0071