Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich HYDROXYAPATITE COATING POWDER ≥95% PB CONTENT≤50 PPM

Catalog No. 92436910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
Catalog No. Quantity
92-436-910G 10 g
1 options

Catalog No. 92-436-910G

Supplier: Sigma Aldrich 92436910G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

HYDROXYAPATITE COATING POWDER ≥95% PB CONTENT =<50PPM

Specifications

CAS 2167-7-4--7
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.