Learn More
TARGETMOL CHEMICALS INC HYDROXYBUPROPION 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T115885MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hydroxybupropion (6-Hydroxybupropion) is the major active metabolite of Bupropion and an antagonist of nACh receptor. Hydroxybupropion inhibits norepinephrine uptake (IC50 = 1.7 uM).
Catalog No. 50-258-1520