Learn More
Apexbio Technology LLC Hydroxychloroquine 118-42-3 250mg
SDP
Catalog No. 5000089639
Encompass_Preferred
Hydroxychloroquine (CAS 118-42-3) is a synthetic oral agent widely utilized in biomedical research related to malaria and autoimmune disorders such as systemic lupus erythematosus and rheumatoid arthritis Mechanistically hydroxychloroquine inhibits autophagic flux by altering lysosomal pH thereby disrupting autophagic degradation pathways It also interferes with Toll-like receptor 7 and 9 (TLR7/9) signaling modulating innate immune responses Additionally the compound exhibits antiviral properties in experimental systems Hydroxychloroquine is used to study immunomodulation autophagy and viral infection mechanisms in cellular and molecular research contexts
Catalog No. 50-000-89639 Supplier Apexbio Technology LLC Supplier No. BA2043250
Hydroxychloroquine (CAS 118-42-3) is a synthetic oral agent widely utilized in biomedical research related to malaria and autoimmune disorders such as systemic lupus erythematosus and rheumatoid arthritis Mechanistically hydroxychloroquine inhibits autophagic flux by altering lysosomal pH thereby disrupting autophagic degradation pathways It also interferes with Toll-like receptor 7 and 9 (TLR7/9) signaling modulating innate immune responses Additionally the compound exhibits antiviral properties in experimental systems Hydroxychloroquine is used to study immunomodulation autophagy and viral infection mechanisms in cellular and molecular research contexts