Learn More
Medchemexpress LLC Hydroxychloroquine | 118-42-3 | MFCD00242707 | 99.8% | 335.88 g/mol | C18H26ClN3O | 250 MG
SDP
Catalog No. 5000303470
Encompass_Preferred
Hydroxychloroquine is a synthetic antimalarial agent widely used in research to study malaria, autoimmune disease pathways, autophagy, and antiviral mechanisms. Supplied as a high-purity solid for laboratory use, it is intended for preparation of stock solutions and as a reference for in vitro assays.
  • High purity suitable for research use.
  • Available in multiple solid pack sizes for flexible experiment planning.
  • Acts as an autophagy inhibitor and modulates TLR7/9 signaling.
  • Compatible with common solvents for preparation of stock solutions.
  • Provided with CAS and analytical data for traceability.
Catalog No. 50-003-03470 Supplier Medchemexpress LLC Supplier No. 250MGHYW031727
Hydroxychloroquine is a synthetic antimalarial agent widely used in research to study malaria, autoimmune disease pathways, autophagy, and antiviral mechanisms. Supplied as a high-purity solid for laboratory use, it is intended for preparation of stock solutions and as a reference for in vitro assays.
  • High purity suitable for research use.
  • Available in multiple solid pack sizes for flexible experiment planning.
  • Acts as an autophagy inhibitor and modulates TLR7/9 signaling.
  • Compatible with common solvents for preparation of stock solutions.
  • Provided with CAS and analytical data for traceability.