Learn More
AdipoGen Hydroxychloroquine . sulfate (250 mg)
SDP
Supplier:  AdipoGen AGCR13720M250
Encompass_Preferred
Chemical. CAS: 747-36-4. Formula: C18H26ClN3O . H2SO4. MW: 335.9 . 98.1. Hydroxychloroquine is a synthetic less toxic derivative of chloroquine (Prod. No. AG-CR1-3721), commonly used as an antimalarial drug. Hydroxychloroquine has anti-inflammatory, immunomodulating, anti-infective, antiviral, antithrombotic and metabolic effects. It has anticancer properties, related to their strong antiproliferative, antimutagenic, epigenetic and autophagy inhibiting and apoptosis inducing activities. It increases lysosomal pH in antigen-presenting cells and blocks in inflammatory conditions, toll-like receptors on plasmacytoid dendritic cells (PDCs). By decreasing TLR signaling, it reduces the activation of dendritic cells and the inflammatory process. It also acts as an antagonist TLR7 and TLR9. Hydroxychloroquine shows antiviral activity against several viruses by inhibiting viral replication and inhibits SARS-CoV-2 viral infection (COVID-19), in vitro.
Catalog No. 50-205-4117