Learn More
Selleck Chemical LLC Hydroxychloroquine Sulfate 10mg 747-36-4 HCQ
SDP
Supplier:  Selleck Chemical LLC S443010MG
Encompass_Preferred
Hydroxychloroquine Sulfate (HCQ) is an antimalarial agent used for the treatment of systemic lupus erythematosus, rheumatoid arthritis and other autoimmune, inflammatory and dermatologic conditions. Also acts as an inhibitor of autophagy and toll-like receptor (TLR) 7/9. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0980049