Learn More
American Radiolabeled Chemicals Inc HYDROXYCHOL 26,27 3H 250UC
SDP
Catalog No. 50353012
Encompass
Hydroxycholesterol 25-26,27-3H 250uCi in ethanol, 1 mCi/ml, 10-30 Ci/mmol, M.W. 402.66, Shipped in Dry Ice
Catalog No. 50-353-012 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0766A250UC
Add to Cart
Add to Cart
Request a Quote
Hydroxycholesterol 25-26,27-3H 250uCi in ethanol, 1 mCi/ml, 10-30 Ci/mmol, M.W. 402.66, Shipped in Dry Ice