Learn More
Medchemexpress LLC HY-16007 5mg , (-)-Hydroxycitric acid CAS:27750-10-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160075MG
Encompass_Preferred
HY-16007 5mg (-)-Hydroxycitric acid CAS: 27750-10-3 (-)-Hydroxycitric acid (Garcinia acid) is the principal acid of fruit rinds of Garcinia cambogia. (-)-Hydroxycitric acid is a potent and competitive inhibitor of ATP citrate lyase. (-)-Hydroxycitric acid suppresses the fatty acid synthesis, lipogenesis, food intake, and induced weight loss. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1489