Learn More
Medchemexpress LLC HY-W009156 500mg Medchemexpress, Hydroxycitric acid (tripotassium hydrate) CAS:6100-05-6 Purity:>98%
SDP
Catalog No. 502083265
Encompass_Preferred
Medchemexpress, HY-W009156 500mg Hydroxycitric acid (tripotassium hydrate) CAS:6100-05-6 Hydroxycitric acid tripotassium hydrate (Potassium citrate monohydrate) is the major active ingredient of Garcinia cambogia and a derivative of citric acid. Hydroxycitric acid tripotassium hydrate competitively inhibits ATP citrate lyase with weight loss benefits. Hydroxycitric acid tripotassium hydrate effective inhibits stones formation and also inhibits HIF, and has antioxidation, anti-inflammation and anti-tumor effects[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3265 Supplier Medchemexpress LLC Supplier No. HYW009156500MG
Medchemexpress, HY-W009156 500mg Hydroxycitric acid (tripotassium hydrate) CAS:6100-05-6 Hydroxycitric acid tripotassium hydrate (Potassium citrate monohydrate) is the major active ingredient of Garcinia cambogia and a derivative of citric acid. Hydroxycitric acid tripotassium hydrate competitively inhibits ATP citrate lyase with weight loss benefits. Hydroxycitric acid tripotassium hydrate effective inhibits stones formation and also inhibits HIF, and has antioxidation, anti-inflammation and anti-tumor effects[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.