Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pfaltz & Bauer HYDROXYCITRONELLAL 95%

Catalog No. 507490104
Encompass

Hydroxycitronellal 95%; 25g; CAS: 107-75-5

Catalog No. 50-749-0104 Supplier Pfaltz & Bauer Supplier No. H1066025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxycitronellal 95%; 25g; CAS: 107-75-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.