Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxydicyclopentadiene (so called) 90.0+%, TCI America™
SDP

Catalog No. H0384500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0384500ML 500 mL
H038425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0384500ML Supplier TCI America Supplier No. H0384500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37275-49-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00151152
InChI Key LDUKQFUHJZHLRC-YZUYTKQYSA-N
Synonym 8(9)-Hydroxytricyclo[5.2.1.0(2,6)]dec-3-ene
PubChem CID 131632538
SMILES C1C=CC2C1C3CC2C(C3)O

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 150.22
Percent Purity ≥90.0% (GC)
Physical Form Viscous Liquid
Chemical Name or Material Hydroxydicyclopentadiene (so called)
EINECSNumber (4)-1229
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.