Learn More
AdipoGen Hydroxyectoine
SDP
Supplier:  AdipoGen AGCN20392G001
Encompass_Preferred
Natural chemical product. CAS 165542-15-4. Formula C6H10N2O3. MW 158.2. Isolated from halophilic bacteria Halomonas elongata. The ectoines belong to the class of compatible solutes also called extremolytes osmolytes from extremophiles. Extremolytes stabilize biological structures and macromolecules such as lipid membranes and proteins against a variety of stress factors, including high temperature, freeze-thaw cycles, drying and degradation. These very stable, non-toxic, animal-free protective agents are powerful additives in various Life Science applications, ranging from cryoprotection of animal cells over protein storage to PCR. They are protective osmolytes selected by nature for stabilization of all proteins in the cell against denaturing environmental stress without altering their functional activity. The stabilizing effect is based on the fact, that the protective osmolytes are strong water-structure formers and as such are preferentially excluded from the protein surface.
Catalog No. 50-256-2556
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.