Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxyethyl disulfide | 1892-29-1 | MFCD00002906 | 98.8% | 154.25 g/mol | C4H10O2S2 | 10 G
SDP

Catalog No. 5000209359
Encompass_Preferred

Hydroxyethyl disulfide is a small-molecule laboratory reagent used in biochemical research. It is reported to interact with thymidylate synthase pathways and is provided with supporting documentation such as a certificate of analysis and safety data sheet. The compound is commonly used as a reagent or intermediate in chemical and biochemical studies.

  • Purity 98.8% (reported)
  • Cas number 1892-29-1
  • Molecular formula C4H10O2S2
  • Molecular weight 154.25 g/mol
  • Available as a 10 G package

Catalog No. 50-002-09359 Supplier Medchemexpress LLC Supplier No. HYW076898A10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxyethyl disulfide is a small-molecule laboratory reagent used in biochemical research. It is reported to interact with thymidylate synthase pathways and is provided with supporting documentation such as a certificate of analysis and safety data sheet. The compound is commonly used as a reagent or intermediate in chemical and biochemical studies.

  • Purity 98.8% (reported)
  • Cas number 1892-29-1
  • Molecular formula C4H10O2S2
  • Molecular weight 154.25 g/mol
  • Available as a 10 G package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.