Learn More
Medchemexpress LLC Hydroxyethyl disulfide | 1892-29-1 | MFCD00002906 | 98.8% | 154.25 g/mol | C4H10O2S2 | 25 G
SDP
Catalog No. 5000208615
Encompass_Preferred
Hydroxyethyl disulfide is a small-molecule disulfide reagent used in biochemical research and enzymology. It has been reported to block the inhibitory effect of showdomycin on methotrexate-resistant thymidylate synthase. Prior to use, consult the safety data sheet and certificate of analysis for handling and quality information.
  • High purity suitable for research use.
  • Single CAS-registered chemical for unambiguous identification.
  • Simple molecular composition with formula C4H10O2S2.
  • Available in multiple sizes, including a 25 g package.
  • Safety and quality documentation (SDS, COA, datasheet) available.
Catalog No. 50-002-08615 Supplier Medchemexpress LLC Supplier No. 25GHYW076898A
Hydroxyethyl disulfide is a small-molecule disulfide reagent used in biochemical research and enzymology. It has been reported to block the inhibitory effect of showdomycin on methotrexate-resistant thymidylate synthase. Prior to use, consult the safety data sheet and certificate of analysis for handling and quality information.
  • High purity suitable for research use.
  • Single CAS-registered chemical for unambiguous identification.
  • Simple molecular composition with formula C4H10O2S2.
  • Available in multiple sizes, including a 25 g package.
  • Safety and quality documentation (SDS, COA, datasheet) available.