Learn More
Crescent Chemical Co Inc HYDROXYETHYL)PIPERAZINE-N'-2-E
SDP
Supplier:  Crescent Chemical Co Inc SE25245.03
Encompass_Preferred
Hydroxyethyl)piperazine-N'-2-ethane sulfonic acid analytical grade, for cell culture pKa 20 = 7.55. Buffering substance (1). Tested for use in tissue culture (2). Physical parameters (3). Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 25245.03 CAS 7365-
Catalog No. 50-247-726