Learn More
Crescent Chemical Co Inc HYDROXYETHYL)PIPERAZINE-N'-2-E
SDP
Catalog No. 50247731
Encompass_Preferred
Hydroxyethyl)piperazine-N'-2-ethane sulfonic acid -Na-salt analytical grade pKa 20 = 7.55. Buffering substance for biochemistry and molecular biology. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 25249.04 CAS 75277-39-3
Catalog No. 50-247-731 Supplier Crescent Chemical Co Inc Supplier No. SE25249.04
Hydroxyethyl)piperazine-N'-2-ethane sulfonic acid -Na-salt analytical grade pKa 20 = 7.55. Buffering substance for biochemistry and molecular biology. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 25249.04 CAS 75277-39-3