Learn More
Cayman Chemical Hydroxyfasudil, Purity: ≥98%, CAS: 105628-72-6, 50 mg
SDP
Catalog No. 502943411
Encompass_Preferred
An inhibitor of ROCK1 and ROCK2 (IC50s = 0.15 and 0.57 µM, respectively) and an active metabolite of HA-1077; decreases blood viscosity and prevents cerebral vasospasm in a canine model of subarachnoid hemorrhage at 3 and 10 mg/kg; reverses hypertension-associated dysfunction of NO-induced relaxation in corpus cavernosum smooth muscle in spontaneously hypertensive rats
Catalog No. 50-294-3411 Supplier Cayman Chemical Supplier No. 4081450 MG
An inhibitor of ROCK1 and ROCK2 (IC50s = 0.15 and 0.57 µM, respectively) and an active metabolite of HA-1077; decreases blood viscosity and prevents cerebral vasospasm in a canine model of subarachnoid hemorrhage at 3 and 10 mg/kg; reverses hypertension-associated dysfunction of NO-induced relaxation in corpus cavernosum smooth muscle in spontaneously hypertensive rats