Learn More
Medchemexpress LLC HY-13911A 5mg Medchemexpress, Hydroxyfasudil (hydrochloride) CAS:155558-32-0 Purity:>98%
SDP
Catalog No. 502030491
Encompass_Preferred
Medchemexpress, HY-13911A 5mg Hydroxyfasudil (hydrochloride) CAS:155558-32-0 Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0491 Supplier Medchemexpress LLC Supplier No. HY13911A5MG
Medchemexpress, HY-13911A 5mg Hydroxyfasudil (hydrochloride) CAS:155558-32-0 Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.