Learn More
Medchemexpress LLC Hydroxyflutamide | 52806-53-8 | 99.77% | 292.21 | 10 MG
SDP
Catalog No. 5000454422
Encompass_Preferred
Hydroxyflutamide (HFT) is the active metabolite of Flutamide and is orally active. It acts as a potent androgen receptor antagonist with an IC50 of 700 nM. This compound can impact embryonic and reproductive tract development in mice, and it also has the potential to enhance the efficacy of Bacillus Calmette-Guérin (BCG) in inhibiting bladder cancer progression. Hydroxyflutamide is used in research related to tumors and reproductive diseases.
  • Potent androgen receptor antagonist
  • Affects embryonic and reproductive tract development
  • Enhances BCG efficacy in bladder cancer research
  • Used in tumor and reproductive disease research
Catalog No. 50-004-54422 Supplier Medchemexpress LLC Supplier No. HYW01327210MG
Hydroxyflutamide (HFT) is the active metabolite of Flutamide and is orally active. It acts as a potent androgen receptor antagonist with an IC50 of 700 nM. This compound can impact embryonic and reproductive tract development in mice, and it also has the potential to enhance the efficacy of Bacillus Calmette-Guérin (BCG) in inhibiting bladder cancer progression. Hydroxyflutamide is used in research related to tumors and reproductive diseases.
  • Potent androgen receptor antagonist
  • Affects embryonic and reproductive tract development
  • Enhances BCG efficacy in bladder cancer research
  • Used in tumor and reproductive disease research