Learn More
Medchemexpress LLC α-Hydroxyglutaric acid-13C5 disodium | 2482467-23-0 | 99.8% | 197.04 | 1 MG
SDP
Catalog No. 5000440045
Encompass_Preferred
α-Hydroxyglutaric acid-13C5 (sodium) is the 13C labeled α-Hydroxyglutaric acid sodium. It is an α-hydroxy acid form of glutaric acid and acts as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methlycytosine (5mC) hydroxylases. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-40045 Supplier Medchemexpress LLC Supplier No. HY113038AS1MG
α-Hydroxyglutaric acid-13C5 (sodium) is the 13C labeled α-Hydroxyglutaric acid sodium. It is an α-hydroxy acid form of glutaric acid and acts as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methlycytosine (5mC) hydroxylases. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.