Learn More
Medchemexpress LLC α-Hydroxyglutaric acid | 2889-31-8 | ≥99.0% | 148.11 | 25 MG
SDP
Catalog No. 5000439421
Encompass_Preferred
α-Hydroxyglutaric acid (2-Hydroxyglutarate) is an α-hydroxy acid form of glutaric acid. It acts as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methylcytosine (5mC) hydroxylases. Isocitrate Dehydrogenase 1 (IDH1) and IDH2 mutations, frequently found in gliomas and acute myeloid leukemia, lead to the production of α-Hydroxyglutaric acid.
  • Inhibits multiple histone demethylases by occupying the α-KG active site.
  • Inhibits TET 5-methylcytosine hydroxylases.
  • Treatment of U-87MG cells with 10-50 mM increases HIF-1α and decreases endostatin.
Catalog No. 50-004-39421 Supplier Medchemexpress LLC Supplier No. 25MGHY113038B
α-Hydroxyglutaric acid (2-Hydroxyglutarate) is an α-hydroxy acid form of glutaric acid. It acts as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methylcytosine (5mC) hydroxylases. Isocitrate Dehydrogenase 1 (IDH1) and IDH2 mutations, frequently found in gliomas and acute myeloid leukemia, lead to the production of α-Hydroxyglutaric acid.
  • Inhibits multiple histone demethylases by occupying the α-KG active site.
  • Inhibits TET 5-methylcytosine hydroxylases.
  • Treatment of U-87MG cells with 10-50 mM increases HIF-1α and decreases endostatin.