Learn More
Medchemexpress LLC α-Hydroxyglutaric acid disodium | 40951-21-1 | 98.0% | 192.08 | 100 MG
SDP
Catalog No. 5000439796
Encompass_Preferred
α-Hydroxyglutaric acid (2-Hydroxyglutarate) disodium is an α-hydroxy acid form of glutaric acid. It is a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methlycytosine (5mC) hydroxylases.
  • Competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases.
  • Inhibits histone demethylases.
  • Inhibits the TET family of 5-methylcytosine (5mC) hydroxylases.
Catalog No. 50-004-39796 Supplier Medchemexpress LLC Supplier No. HY113038A100MG
α-Hydroxyglutaric acid (2-Hydroxyglutarate) disodium is an α-hydroxy acid form of glutaric acid. It is a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methlycytosine (5mC) hydroxylases.
  • Competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases.
  • Inhibits histone demethylases.
  • Inhibits the TET family of 5-methylcytosine (5mC) hydroxylases.