Learn More
Medchemexpress LLC α-Hydroxyglutaric acid disodium | 40951-21-1 | 98.0% | 192.08 | 25 MG
SDP
Catalog No. 5000440008
Encompass_Preferred
α-Hydroxyglutaric acid disodium is an α-hydroxy acid form of glutaric acid. It is a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methylcytosine (5mC) hydroxylases. Research indicates that isocitrate dehydrogenase 1 (IDH1) and IDH2 mutations, common in gliomas and acute myeloid leukemia, lead to altered production of α-ketoglutarate and α-Hydroxyglutaric acid disodium. Furthermore, it has been shown to inhibit the activity of TET 5-methylcytosine hydroxylases and, in U-87MG cells, increases HIF-1α while decreasing endostatin.
  • Endogenous metabolite
  • Competitive inhibitor of α-ketoglutarate-dependent dioxygenases
  • Inhibits histone demethylases
  • Inhibits TET family of 5-methylcytosine (5mC) hydroxylases
  • Increases HIF-1α and decreases endostatin in U-87MG cells
Catalog No. 50-004-40008 Supplier Medchemexpress LLC Supplier No. 25MGHY113038A
α-Hydroxyglutaric acid disodium is an α-hydroxy acid form of glutaric acid. It is a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases, including histone demethylases and the TET family of 5-methylcytosine (5mC) hydroxylases. Research indicates that isocitrate dehydrogenase 1 (IDH1) and IDH2 mutations, common in gliomas and acute myeloid leukemia, lead to altered production of α-ketoglutarate and α-Hydroxyglutaric acid disodium. Furthermore, it has been shown to inhibit the activity of TET 5-methylcytosine hydroxylases and, in U-87MG cells, increases HIF-1α while decreasing endostatin.
  • Endogenous metabolite
  • Competitive inhibitor of α-ketoglutarate-dependent dioxygenases
  • Inhibits histone demethylases
  • Inhibits TET family of 5-methylcytosine (5mC) hydroxylases
  • Increases HIF-1α and decreases endostatin in U-87MG cells