Learn More
Medchemexpress LLC α-hydroxyglutaric acid (disodium) | 40951-21-1 | 98.0% | 192.08 g·mol⁻1 | C5H6Na2O5 | 5 MG
SDP
Catalog No. 5000262480
Encompass_Preferred
α-Hydroxyglutaric acid disodium is the disodium salt form of α-hydroxyglutaric acid supplied for biochemical research. It functions as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases and is commonly used in studies of epigenetic regulation and metabolite signaling. The material is offered at a high stated purity and in small milligram pack sizes suitable for assay-scale experiments.
  • Competitive inhibitor of α-ketoglutarate-dependent dioxygenases.
  • Suitable for epigenetic and metabolic pathway studies.
  • Supplied as disodium salt for improved aqueous solubility.
  • High stated purity for reproducible experimental results.
  • Available in small milligram quantities for assay-scale use.
Catalog No. 50-002-62480 Supplier Medchemexpress LLC Supplier No. HY113038A5MG
α-Hydroxyglutaric acid disodium is the disodium salt form of α-hydroxyglutaric acid supplied for biochemical research. It functions as a competitive inhibitor of multiple α-ketoglutarate-dependent dioxygenases and is commonly used in studies of epigenetic regulation and metabolite signaling. The material is offered at a high stated purity and in small milligram pack sizes suitable for assay-scale experiments.
  • Competitive inhibitor of α-ketoglutarate-dependent dioxygenases.
  • Suitable for epigenetic and metabolic pathway studies.
  • Supplied as disodium salt for improved aqueous solubility.
  • High stated purity for reproducible experimental results.
  • Available in small milligram quantities for assay-scale use.