Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyguanidine Sulfate, Hydrate 99% (on the Dry Basis), Thermo Scientific™

Catalog No. AC412030250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412030250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412030250 Supplier Thermo Scientific Chemicals Supplier No. 412030250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6345-29-5
Molecular Formula C2H10N6O2.H2SO4.xH2O

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula (NH2C(NH)NHOH)2.H2SO4.xH2O
Quantity 25 g
Formula Weight 248.21
Percent Purity ≥98.5% (on dry substance)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxyguanidine sulfate, hydrate, 99% (on the dry basis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.