Learn More
Selleck Chemical LLC Hydroxyhexamide
SDP
Catalog No. 5000002870
Encompass_Preferred
Hydroxyhexamide is a pharmacologically active metabolite of acetohexamide which is a sulfonylurea hypoglycemic agent
Catalog No. 50-000-02870 Supplier Selleck Chemical LLC Supplier No. S56722MG
Hydroxyhexamide is a pharmacologically active metabolite of acetohexamide which is a sulfonylurea hypoglycemic agent