Learn More
Medchemexpress LLC Hydroxyisohexyl 3-cyclohexene carboxaldehyde | 31906-04-4 | MFCD00019423 | 97.8% | 210.31 | C13H22O2 | 50 MG
SDP
Catalog No. 5000299846
Encompass_Preferred
Hydroxyisohexyl 3-cyclohexene carboxaldehyde is a fragrance aldehyde used as a hapten probe in research. Supplied as a colorless to light yellow liquid with supporting analytical documentation, it is intended for use in biochemical assays, allergen testing, and analytical studies where a well-characterized fragrance hapten is required.
  • Used as a hapten probe in fragrance and contact allergy research.
  • High purity (97.8%) suitable for analytical use.
  • Molecular formula C13H22O2, molecular weight 210.31.
  • Available as neat material or as a 10 mM solution in DMSO for biological assays.
  • Documentation provided: COA, SDS, HNMR, GC for quality verification.
Catalog No. 50-002-99846 Supplier Medchemexpress LLC Supplier No. HYW03521450MG
Hydroxyisohexyl 3-cyclohexene carboxaldehyde is a fragrance aldehyde used as a hapten probe in research. Supplied as a colorless to light yellow liquid with supporting analytical documentation, it is intended for use in biochemical assays, allergen testing, and analytical studies where a well-characterized fragrance hapten is required.
  • Used as a hapten probe in fragrance and contact allergy research.
  • High purity (97.8%) suitable for analytical use.
  • Molecular formula C13H22O2, molecular weight 210.31.
  • Available as neat material or as a 10 mM solution in DMSO for biological assays.
  • Documentation provided: COA, SDS, HNMR, GC for quality verification.