Learn More
Medchemexpress LLC Hydroxyisohexyl 3-cyclohexene carboxaldehyde | 31906-04-4 | MFCD00019423 | 97.8% | 210.31 g/mol | C13H22O2 | 1 ML
SDP
Catalog No. 5000299353
Encompass_Preferred
Hydroxyisohexyl 3-cyclohexene carboxaldehyde (CAS 31906-04-4) is a fragrance hapten probe provided for laboratory research, available as a liquid or as a 10 mM solution in DMSO. The material is high-purity (≈97.8% by GC), has formula C13H22O2 and molar mass 210.31 g/mol, and is supplied colorless to light yellow; store under nitrogen and follow recommended temperature conditions.
Catalog No. 50-002-99353 Supplier Medchemexpress LLC Supplier No. HYW03521410MM/1ML
Hydroxyisohexyl 3-cyclohexene carboxaldehyde (CAS 31906-04-4) is a fragrance hapten probe provided for laboratory research, available as a liquid or as a 10 mM solution in DMSO. The material is high-purity (≈97.8% by GC), has formula C13H22O2 and molar mass 210.31 g/mol, and is supplied colorless to light yellow; store under nitrogen and follow recommended temperature conditions.