Learn More
Cayman Chemical Hydroxyisovaleric ACD 10g
SDP
Catalog No. 502950308
Encompass_Preferred
An active metabolite of L-leucine; reduces PIF-induced increases in 20S proteasome activity and NF-κB nuclear accumulation in C2C12 myotubes; inhibits protein degradation and stimulates protein synthesis in the gastrocnemius muscle of cachectic mice bearing MAC16 tumor at 0.25 and 2.5 g/kg; urinary levels are increased in patients with type II diabetes or ketoacidosis
Catalog No. 50-295-0308 Supplier Cayman Chemical Supplier No. 3403010 G
An active metabolite of L-leucine; reduces PIF-induced increases in 20S proteasome activity and NF-κB nuclear accumulation in C2C12 myotubes; inhibits protein degradation and stimulates protein synthesis in the gastrocnemius muscle of cachectic mice bearing MAC16 tumor at 0.25 and 2.5 g/kg; urinary levels are increased in patients with type II diabetes or ketoacidosis