Learn More
Medchemexpress LLC Hydroxylamine-d3 hydrochloride | 15588-23-5 | MFCD00075189 | 99.0% | 72.51 g/mol | ClHD3NO | 50mg
SDP
Catalog No. 5000268521
Encompass_Preferred
Hydroxylamine-d3 (hydrochloride) is the deuterium labeled Hydroxyamine hydrochloride Hydroxylamine hydrochloride is a selective Monoamine oxidase (MAO) inhibitor used for inhibiting of platelet aggregation Hydroxylamine hydrochloride is an intermediate of nitrogen cycle in aerobic ammonium oxidizers microorganisms such as ammonium oxidizing bacteria (AOB) ammonium oxidizing archaea (AOA) and complete ammonium oxidizing bacteria (comammox) Hydroxylamine hydrochloride impacts NO and N2O emissions by aerobic ammonium oxidizers microorganisms and inhibits nitrite oxidizing bacteria (NOB) activity[1][2][3]
Catalog No. 50-002-68521 Supplier Medchemexpress LLC Supplier No. HYY0882S150MG
Hydroxylamine-d3 (hydrochloride) is the deuterium labeled Hydroxyamine hydrochloride Hydroxylamine hydrochloride is a selective Monoamine oxidase (MAO) inhibitor used for inhibiting of platelet aggregation Hydroxylamine hydrochloride is an intermediate of nitrogen cycle in aerobic ammonium oxidizers microorganisms such as ammonium oxidizing bacteria (AOB) ammonium oxidizing archaea (AOA) and complete ammonium oxidizing bacteria (comammox) Hydroxylamine hydrochloride impacts NO and N2O emissions by aerobic ammonium oxidizers microorganisms and inhibits nitrite oxidizing bacteria (NOB) activity[1][2][3]