Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine-EDTA, For Selenium, LabChem™

Catalog No. LC155101 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC155101 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC155101 Supplier LabChem, Inc. Supplier No. LC155101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Hydroxylamine-EDTA
Color Colorless
Name Note For Selenium
CAS Max % 96.6, 2.5, 0.9
CAS 5470-11-1, 6381-92-6
Linear Formula H3NO·HCl
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 69.49
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.