Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydroxylamine hydrochlorid
Catalog No. 502897771
Encompass
Hydroxylamine hydrochloride (Hydroxylammonium chloride) participates in the synthesis of 1 2 4-oxadiazoles secondary amides and tertiary amides.
Catalog No. 50-289-7771 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1594173KG
Hydroxylamine hydrochloride (Hydroxylammonium chloride) participates in the synthesis of 1 2 4-oxadiazoles secondary amides and tertiary amides.