Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydroxylamine hydrochlorid1KG
Catalog No. 502898904
Encompass
Hydroxylamine hydrochloride (Hydroxylammonium chloride) is the hydrochloride salt of hydroxylamine. It participates in the synthesis of 1 2 4-oxadiazoles.
Catalog No. 50-289-8904 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 554601KG
Hydroxylamine hydrochloride (Hydroxylammonium chloride) is the hydrochloride salt of hydroxylamine. It participates in the synthesis of 1 2 4-oxadiazoles.