Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hydroxylamine Hydrochloride, 113 g

Catalog No. 501994216 Shop All Hach Company Products
Encompass

ACS Grade, CAS No. 5470-11-1 NH2OH.HCl, FW = 69.49 Assay ≥96.0%

Catalog No. 50-199-4216 Supplier Hach Company Supplier No. 24614
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

ACS Grade, CAS No. 5470-11-1 NH2OH.HCl, FW = 69.49 Assay ≥96.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.