Learn More
Medchemexpress LLC Hydroxylamine hydrochloride | 5470-11-1 | MFCD00051089 | 99.9% | 69.49 g·mol⁻¹ | NH2OH · HCl | 5g
SDP
Catalog No. 5000270008
Encompass_Preferred
Hydroxylamine hydrochloride is a selective Monoamine oxidase (MAO) inhibitor used for inhibiting of platelet aggregation Hydroxylamine hydrochloride is an intermediate of nitrogen cycle in aerobic ammonium oxidizers microorganisms such as ammonium oxidizing bacteria (AOB) ammonium oxidizing archaea (AOA) and complete ammonium oxidizing bacteria (comammox) Hydroxylamine hydrochloride impacts NO and N2O emissions by aerobic ammonium oxidizers microorganisms and inhibits nitrite oxidizing bacteria (NOB) activity[1][2]
Catalog No. 50-002-70008 Supplier Medchemexpress LLC Supplier No. HYY08825G
Hydroxylamine hydrochloride is a selective Monoamine oxidase (MAO) inhibitor used for inhibiting of platelet aggregation Hydroxylamine hydrochloride is an intermediate of nitrogen cycle in aerobic ammonium oxidizers microorganisms such as ammonium oxidizing bacteria (AOB) ammonium oxidizing archaea (AOA) and complete ammonium oxidizing bacteria (comammox) Hydroxylamine hydrochloride impacts NO and N2O emissions by aerobic ammonium oxidizers microorganisms and inhibits nitrite oxidizing bacteria (NOB) activity[1][2]