Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxylamine, o-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]- | 1306615-51-9 | 95.0% | 234.25 | C8H18N4O4 | 25 MG
SDP

Catalog No. 5000202345
Encompass_Preferred

Aminooxy-PEG3-azide is a bifunctional PEG-based linker bearing an aminooxy group and an azide group, suitable for conjugation chemistry. It serves as a non-cleavable three-unit PEG spacer for synthesis of antibody-drug conjugates and PROTACs, and enables click chemistry reactions such as CuAAC and SPAAC.

  • Used as a non-cleavable three-unit PEG linker for conjugation chemistry
  • Contains aminooxy and azide functional groups for orthogonal conjugation
  • Compatible with CuAAC and SPAAC click reactions
  • Molecular weight 234.25 and formula C8H18N4O4
  • Typical purity 95.0% (NMR)

Catalog No. 50-002-02345 Supplier Medchemexpress LLC Supplier No. HY12694925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminooxy-PEG3-azide is a bifunctional PEG-based linker bearing an aminooxy group and an azide group, suitable for conjugation chemistry. It serves as a non-cleavable three-unit PEG spacer for synthesis of antibody-drug conjugates and PROTACs, and enables click chemistry reactions such as CuAAC and SPAAC.

  • Used as a non-cleavable three-unit PEG linker for conjugation chemistry
  • Contains aminooxy and azide functional groups for orthogonal conjugation
  • Compatible with CuAAC and SPAAC click reactions
  • Molecular weight 234.25 and formula C8H18N4O4
  • Typical purity 95.0% (NMR)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.