Learn More
Medchemexpress LLC Hydroxylamine, O-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl] | 1306615-51-9 | 95.0% | 234.25 g/mol | C8H18N4O4 | 100 MG
SDP
Catalog No. 5000202344
Encompass_Preferred
Aminooxy-PEG3-azide is a PEG-based bifunctional linker containing an aminooxy (hydroxylamine) group and an azide group for orthogonal bioconjugation. It enables oxime formation with aldehydes or ketones and azide-alkyne click chemistry, and is supplied as a liquid at high purity for lab-scale synthesis.
  • Contains aminooxy and azide functional groups for orthogonal conjugation
  • Enables oxime ligation and azide-alkyne click reactions
  • Suitable for ADC, PROTAC, and general bioconjugation workflows
  • Supplied as a liquid with high laboratory-grade purity
  • Available in small-scale pack sizes for research use
Catalog No. 50-002-02344 Supplier Medchemexpress LLC Supplier No. HY126949100MG
Aminooxy-PEG3-azide is a PEG-based bifunctional linker containing an aminooxy (hydroxylamine) group and an azide group for orthogonal bioconjugation. It enables oxime formation with aldehydes or ketones and azide-alkyne click chemistry, and is supplied as a liquid at high purity for lab-scale synthesis.
  • Contains aminooxy and azide functional groups for orthogonal conjugation
  • Enables oxime ligation and azide-alkyne click reactions
  • Suitable for ADC, PROTAC, and general bioconjugation workflows
  • Supplied as a liquid with high laboratory-grade purity
  • Available in small-scale pack sizes for research use