Learn More
Medchemexpress LLC Hydroxylamine, O-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]- | 1306615-51-9 | 95.0% | 234.25 g/mol | C8H18N4O4 | 5 MG
SDP
Catalog No. 5000202343
Encompass_Preferred
Aminooxy-PEG3-azide is a bifunctional PEG linker containing an aminooxy group and an azide handle for oxime ligation and azide-alkyne click chemistry. It is supplied as a liquid and is used for ADC and PROTAC linker synthesis, and for general bioconjugation and small-molecule assembly.
  • Bifunctional: aminooxy and azide reactive groups.
  • Peg3 spacer provides hydrophilicity and flexibility.
  • Suitable for oxime formation with carbonyls and CuAAC click reactions.
  • Purity typically 95.0%.
  • Stable when stored at -20°C; in solvent short-term storage at lower temperatures recommended.
  • Available in small research quantities, such as 5 mg.
Catalog No. 50-002-02343 Supplier Medchemexpress LLC Supplier No. HY1269495MG
Aminooxy-PEG3-azide is a bifunctional PEG linker containing an aminooxy group and an azide handle for oxime ligation and azide-alkyne click chemistry. It is supplied as a liquid and is used for ADC and PROTAC linker synthesis, and for general bioconjugation and small-molecule assembly.
  • Bifunctional: aminooxy and azide reactive groups.
  • Peg3 spacer provides hydrophilicity and flexibility.
  • Suitable for oxime formation with carbonyls and CuAAC click reactions.
  • Purity typically 95.0%.
  • Stable when stored at -20°C; in solvent short-term storage at lower temperatures recommended.
  • Available in small research quantities, such as 5 mg.