Learn More
Medchemexpress LLC Hydroxylamine, O-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]- | 1306615-51-9 | 95.0% | 234.25 g/mol | C8H18N4O4 | 50 MG
SDP
Catalog No. 5000204191
Encompass_Preferred
Aminooxy-PEG3-azide is a bifunctional PEG3 linker reagent containing an aminooxy group and an azide group, used for bioconjugation in ADC and PROTAC synthesis and for click chemistry conjugations.
  • Contains an aminooxy group for oxime ligation.
  • Contains an azide group for CuAAC and SPAAC click chemistry.
  • Three-unit PEG spacer provides solubility and flexibility.
  • High purity suitable for research applications.
  • Stable when stored at -20°C in pure form.
Catalog No. 50-002-04191 Supplier Medchemexpress LLC Supplier No. HY12694950MG
Aminooxy-PEG3-azide is a bifunctional PEG3 linker reagent containing an aminooxy group and an azide group, used for bioconjugation in ADC and PROTAC synthesis and for click chemistry conjugations.
  • Contains an aminooxy group for oxime ligation.
  • Contains an azide group for CuAAC and SPAAC click chemistry.
  • Three-unit PEG spacer provides solubility and flexibility.
  • High purity suitable for research applications.
  • Stable when stored at -20°C in pure form.