Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxylamine, O-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]- | 1306615-51-9 | MFCD28505530 | 95.0% | 234.25 g/mol | C8H18N4O4 | 10 MG
SDP

Catalog No. 5000202088
Encompass_Preferred

Aminooxy-PEG3-azide is a PEG-based bifunctional linker containing an aminooxy (oxyamine) group and an azide group. It is used as a non-cleavable three-unit PEG spacer for antibody-drug conjugate (ADC) and PROTAC synthesis, and as an azide reagent for click chemistry reactions such as CuAAC and SPAAC.

  • Bifunctional aminooxy and azide groups for oxime ligation and click chemistry.
  • Non-cleavable three-unit PEG spacer suitable for ADC and PROTAC linker installation.
  • Purity 95.0% and molecular weight 234.25 g/mol.
  • Appearance colorless to light yellow liquid for easy handling.
  • Storage: pure form -20°C (stable 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes, including 10 mg for small-scale synthesis.

Catalog No. 50-002-02088 Supplier Medchemexpress LLC Supplier No. HY12694910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminooxy-PEG3-azide is a PEG-based bifunctional linker containing an aminooxy (oxyamine) group and an azide group. It is used as a non-cleavable three-unit PEG spacer for antibody-drug conjugate (ADC) and PROTAC synthesis, and as an azide reagent for click chemistry reactions such as CuAAC and SPAAC.

  • Bifunctional aminooxy and azide groups for oxime ligation and click chemistry.
  • Non-cleavable three-unit PEG spacer suitable for ADC and PROTAC linker installation.
  • Purity 95.0% and molecular weight 234.25 g/mol.
  • Appearance colorless to light yellow liquid for easy handling.
  • Storage: pure form -20°C (stable 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes, including 10 mg for small-scale synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.