Learn More
Medchemexpress LLC Hydroxylamine, O-methyl-, hydrochloride (1:1) | 593-56-6 | MFCD00012951 | 98.0% | 83.52 | CH6ClNO | 50g
SDP
Catalog No. 5000268143
Encompass_Preferred
Methoxyamine (O-Methylhydroxylamine) hydrochloride is an orally active and potent base excision repair (BER) inhibitor Methoxyamine hydrochloride binds to 3 hydroxyl groups that are left behind by 3-methylpurine-DNA glycosylase (MPG) following excision of the damaged base and thus inhibits BER activity Methoxyamine hydrochloride binds directly to the apyrimidinic (AP) sites Methoxyamine hydrochloride synergistically enhances the therapeutic efficacy of DNA-damaging agents[1][2]
Catalog No. 50-002-68143 Supplier Medchemexpress LLC Supplier No. HYY095850G
Methoxyamine (O-Methylhydroxylamine) hydrochloride is an orally active and potent base excision repair (BER) inhibitor Methoxyamine hydrochloride binds to 3 hydroxyl groups that are left behind by 3-methylpurine-DNA glycosylase (MPG) following excision of the damaged base and thus inhibits BER activity Methoxyamine hydrochloride binds directly to the apyrimidinic (AP) sites Methoxyamine hydrochloride synergistically enhances the therapeutic efficacy of DNA-damaging agents[1][2]