Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Sulfate, 3% (w/v), Ricca Chemical
SDP

Catalog No. 392732 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3927-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3927-32 Supplier Ricca Chemical Company Supplier No. 392732

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5 , 10039-54-0

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Percent Purity 3% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 3% (w/v)
Odor Odorless
Packaging Natural Poly Bottle
Chemical Name or Material Hydroxylamine Sulfate
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.