Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine sulfate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
5kg
Packaging:
Glass Bottle
Plastic bottle
10039-54-0
H6N2O2·H2SO4
164.14
MFCD00044869
Hydroxylammonium sulfate
10039-54-0
H6N2O2·H2SO4
164.14
MFCD00044869
Hydroxylammonium sulfate
Quantity 1kg
Melting Point 170.0°C
Color White
Packaging Plastic bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (H2NOH)2·H2SO4
Fieser 01,481
Merck Index 15, 4867
Solubility Information Solubility in water: 329g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 164.14
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material Hydroxylamine sulfate
Show More Show Less

GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing ca

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning

EINECSNumber : 233-118-8