Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem Hydroxylapatite, Fast Flow

Catalog No. 391947100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-194-7100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-194-7100GM Supplier MilliporeSigma™ Supplier No. 391947100GM
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A form of calcium phosphate used in preparative procedures for purification of proteins and nucleic acids in which high flow rate and binding capacity are desired. BSA binding capacity: >12mg/g; DNA binding capacity: >700μg/g

  • Formula Weight: 1004.6g/mol
  • Following reconstitution, store in the refrigerator (4°C)
  • Form: White solid
  • CAS Number: 1306-06-5
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.