Learn More
Medchemexpress LLC α-Hydroxylinoleic acid | 57818-44-7 | MFCD31544382 | 296.44 | 50 MG
SDP
Catalog No. 5000448498
Encompass_Preferred
ABTL-0812 (α-Hydroxylinoleic acid) is a first-in-class small molecule that induces endoplasmic reticulum (ER) stress-mediated autophagy, demonstrating anti-cancer activity. It has been observed to inhibit cell viability in squamous NSCLC H157 cells and induce ER stress in human lung and pancreatic xenografts during in vivo studies.
  • Induces ER stress-mediated autophagy
  • Exhibits anti-cancer activity against squamous NSCLC H157 cells
  • Induces ER stress in human lung and pancreatic xenografts
  • First-in-class small molecule
  • Liquid appearance, colorless to off-white
  • Derived from Bombacaceae Pachira aquatica Aublet plants
  • Potential anti-cancer therapeutic
Catalog No. 50-004-48498 Supplier Medchemexpress LLC Supplier No. HYU0014150MG
ABTL-0812 (α-Hydroxylinoleic acid) is a first-in-class small molecule that induces endoplasmic reticulum (ER) stress-mediated autophagy, demonstrating anti-cancer activity. It has been observed to inhibit cell viability in squamous NSCLC H157 cells and induce ER stress in human lung and pancreatic xenografts during in vivo studies.
  • Induces ER stress-mediated autophagy
  • Exhibits anti-cancer activity against squamous NSCLC H157 cells
  • Induces ER stress in human lung and pancreatic xenografts
  • First-in-class small molecule
  • Liquid appearance, colorless to off-white
  • Derived from Bombacaceae Pachira aquatica Aublet plants
  • Potential anti-cancer therapeutic