Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxymethyl dasatinib | 910297-58-4 | MFCD18252610 | 98.0% | 504.00 g/mol | C22H26ClN7O3S | 5 MG
SDP

Catalog No. 5000284307
Encompass_Preferred

Hydroxymethyl dasatinib is the M24 benzylhydroxy metabolite of dasatinib and is supplied as a small-molecule research reagent for in vitro studies. It has been reported to inhibit proliferation of K562 chronic myeloid leukemia cells with an IC50 of 46.7 nM. This compound is intended for research use only and not for human or clinical applications.

  • Benzylhydroxy metabolite of dasatinib.
  • Reported IC50 of 46.7 nM in K562 cells.
  • Small-molecule research reagent for in vitro studies.
  • Molecular weight 504.00 g/mol.
  • Chemical formula C22H26ClN7O3S.
  • CAS number 910297-58-4.
  • Available as 5 MG package.

Catalog No. 50-002-84307 Supplier Medchemexpress LLC Supplier No. HYW7006205MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxymethyl dasatinib is the M24 benzylhydroxy metabolite of dasatinib and is supplied as a small-molecule research reagent for in vitro studies. It has been reported to inhibit proliferation of K562 chronic myeloid leukemia cells with an IC50 of 46.7 nM. This compound is intended for research use only and not for human or clinical applications.

  • Benzylhydroxy metabolite of dasatinib.
  • Reported IC50 of 46.7 nM in K562 cells.
  • Small-molecule research reagent for in vitro studies.
  • Molecular weight 504.00 g/mol.
  • Chemical formula C22H26ClN7O3S.
  • CAS number 910297-58-4.
  • Available as 5 MG package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.