Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxymethyl dasatinib (M24) | 910297-58-4 | >98.0% | 500.56 g/mol | C22H26ClN7O3S | 10 MG
SDP

Catalog No. 5000284942
Encompass_Preferred

Hydroxymethyl dasatinib (M24) is a benzylhydroxy metabolite of dasatinib with reported kinase inhibitory activity. It is supplied as a solid research reagent for in vitro studies of Bcr-Abl and Src-family kinase signaling.

  • High purity >98.0%.
  • Molecular formula C22H26ClN7O3S.
  • CAS number 910297-58-4.
  • Molecular weight 500.56 g/mol.
  • Solid powder form for laboratory research use only.
  • Store powder at -20°C for long-term storage; in solution store at -80°C for short-term use.

Catalog No. 50-002-84942 Supplier Medchemexpress LLC Supplier No. HYW70062010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxymethyl dasatinib (M24) is a benzylhydroxy metabolite of dasatinib with reported kinase inhibitory activity. It is supplied as a solid research reagent for in vitro studies of Bcr-Abl and Src-family kinase signaling.

  • High purity >98.0%.
  • Molecular formula C22H26ClN7O3S.
  • CAS number 910297-58-4.
  • Molecular weight 500.56 g/mol.
  • Solid powder form for laboratory research use only.
  • Store powder at -20°C for long-term storage; in solution store at -80°C for short-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.